Safety funnels with lid, 'MARCO' V2.0
Toimittaja: S.C.A.T.
318951 318955 318950 318954 318952 318953 318990 318991 318992 318993 318994 318995
221-0315EA 121 EUR
221-0315 SCTV318955 SCTV318950 SCTV318954 SCTV318952 SCTV318953 SCTV318990 SCTV318991 SCTV318992 SCTV318993 SCTV318994 SCTV318995
Safety funnels with lid, 'MARCO' V2.0
Tulpat ja korkit Turvakorkit
S.C.A.T. 'MARCO' V2.0 safety funnels can be closed safely and prevent danger of ignition.
  • 'MARCO' V2.0 safety lid funnel is a modern lid funnel made of high quality HDPE with extremely high resistance to acids, alkalis and many chemicals
  • Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents explosive or health-endangering gases escaping
  • Sturdy hinge guarantees the closing mechanism has a long service life