Safety Caps for PFAS Analytic
Toimittaja: S.C.A.T.
307019-PP 307410-PP 307909-PP 307910-PP 397008-PP 397010-PP 399200-PP 399201-PP 399202-PP 399203-PP
SCTV307019-PPEA 148 EUR
SCTV307019-PP SCTV307410-PP SCTV307909-PP SCTV307910-PP SCTV397008-PP SCTV397010-PP SCTV399200-PP SCTV399201-PP SCTV399202-PP SCTV399203-PP
Safety Caps for PFAS Analytic
Tulpat ja korkit Turvakorkit
Safety Caps for PFAS Analytic feature a polypropylene core, which is free from PFAS to ensure the accuracy of PFAS testing without interference from the cap material. All associated accessories such as fittings, blind plugs, and ventilation valves are also PFAS-free.
  • SCAT Safety Caps for PFAS Analytic are hermetically sealing closures for the safe supply of HPLC with solvents
  • Caps are designed for use in tests that aim to detect the presence of fluorine in a substance
  • Caps block harmful vapours and protect HPLC systems from contamination
  • SCAT Air Valves