Flat Flange Lids for Glass Reactors
Toimittaja: S.C.A.T.
130060 130061 130100 130101 130102 130150 130151 130152
SCTV130060EA 193 EUR
SCTV130060 SCTV130061 SCTV130100 SCTV130101 SCTV130102 SCTV130150 SCTV130151 SCTV130152
Flat Flange Lids for Glass Reactors
Kannet Reaction Vessel Lids
The flat flange lid is usually placed between two flanges to be joined and then tightened with bolts to create a secure connection. Suitable for glass reactors with a SCHOTT flat flange.
  • Flat flange allows the lid to be securely attached to the reactor body and ensures a reliable seal to prevent leakage
  • Compatible according to DIN 12214 (laboratory flat flange) and compatible with glass reactors for seamless integration
  • Effortless and hassle-free to assemble, saving time and effort
  • Flexibly expand products to fit specific needs and meet future needs
  • Made
More Product Information
Order Now
TEKNISET TIEDOT
Environmentally Preferable