Exhaust filters for barrels
Toimittaja: S.C.A.T.
108986 108987 108985
SCTV108986EA 567 EUR
SCTV108986 SCTV108987 SCTV108985
Exhaust filters for barrels
Suodattimet Ilmansuodattimet
SCAT exhaust air filters for drums / barrels offer optimal protection against vapors and gases that can escape through opened drums. The high-quality activated carbon prevents clumping when in contact with aerosols and thus prevents the escape of toxic impurities and reliably binds volatile substances.
  • Reliable protection of breathing air in closed rooms and thus the health of employees
  • Guarantee highest quality and safety standards for laboratories and the handling of highly volatile chemicals
  • Contain a high-quality activated carbon granulate, which has