Politainer
Toimittaja: S.C.A.T.
107331 107333 107330 107332
SCTV107331EA 16.9 EUR
SCTV107331 SCTV107333 SCTV107330 SCTV107332
Politainer
Turvakannut
The SCAT Politainer - also known as folding canisters - is a combination of a flexible plastic bag and a stable UN-approved outer carton. Storage in the unfilled state is possible in a very small space and after filling, safe storage or transport is guaranteed.
  • Consists of LDPE and EVA copolymer, which offers optimum protection against volatile solvents
  • Available with the frequently used threads GL38 and S60/61 and with volumes of 5, 10 and 20 L
SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces.