DURAN® Safety Joints, Threaded Socket
Toimittaja: DWK Life Sciences
286581909 286582408 286582905 286584503 286581452 286583404
SCOT286581909EA 169 EUR
SCOT286581909 SCOT286582408 SCOT286582905 SCOT286584503 SCOT286581452 SCOT286583404
DURAN® Safety Joints, Threaded Socket
Sovittimet Sovittimet, NS-hios
NS joints with threaded socket, solid.
  • Made from Borosilicate 3.3 glass
  • Increased wall thickness
  • Threaded sleeve prevents unintentional loosening of the ground joint
  • Firmly seated ground joints can be loosened more easily here, the danger of glass breakage and associated risks is thus minimised
  • Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range
  • Tight closure due to standard ground joint
In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences,More Product Information