Long ground sockets and cones, finished, DURAN®
Toimittaja: DWK Life Sciences
286154602 286155804 286156603 286159206 286162602 286534404 286541605 286546209 286558408
SCOT286154602EA 248 EUR
SCOT286154602 SCOT286155804 SCOT286156603 SCOT286159206 SCOT286162602 SCOT286534404 SCOT286541605 SCOT286546209 SCOT286558408
Long ground sockets and cones, finished, DURAN®
Sovittimet Sovittimet, NS-hios
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes