Sockets and cones, RODAVISS®, DURAN®
Toimittaja: DWK Life Sciences
286214305 286214408 286214502 286214605 286214708 286215001 286215207 286215404 286215507 286215704 286216409 286216606 286216709 286216803 286216906 286217002 286217105 286217208 286217302 286217405 286217508 286217705 286214202 286215301
SCOT286214305EA 47.8 EUR
SCOT286214305 SCOT286214408 SCOT286214502 SCOT286214605 SCOT286214708 SCOT286215001 SCOT286215207 SCOT286215404 SCOT286215507 SCOT286215704 SCOT286216409 SCOT286216606 SCOT286216709 SCOT286216803 SCOT286216906 SCOT286217002 SCOT286217105 SCOT286217208 SCOT286217302 SCOT286217405 SCOT286217508 SCOT286217705 SCOT286214202 SCOT286215301
Sockets and cones, RODAVISS®, DURAN®
Sovittimet Sovittimet, NS-hios
DURAN®, borosilicate glass 3.3, clear.
  • Suitable for work performed under vacuum and pressure
  • Can be used up to a temperature of 200 °C
  • Minimum breakage risk due to anti-jamming system
  • Replaceable with ground joints in accordance with DIN 12 249
The advantages of a glass/glass connection optimised by a securing system. The connections remain absolutely stable and can be easily separated again thanks to the use of components that are technically matched to each other. The connection comprises cone and socket as well as screw cap, safeguarding andMore Product Information