Putket, Attachable screw thread tubes, SVL, DURAN®
Toimittaja: DWK Life Sciences
286064408 286265006 286266208 286266405
SCOT286064408EA 173 EUR
SCOT286064408 SCOT286265006 SCOT286266208 SCOT286266405
Putket, Attachable screw thread tubes, SVL, DURAN®
Sovittimet Adapters, Screwthread tubes
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread SVL. It is used by glassblowers in the construction of laboratory glassware.