Sovittimet, joissa putkiliitin, DURAN®
Toimittaja: DWK Life Sciences
243000604 243000801 243000904
SCOT243000604EA 16.8 EUR
SCOT243000604 SCOT243000801 SCOT243000904
Sovittimet, joissa putkiliitin, DURAN®
Sovittimet Putkistoliittimet, NS-hios
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapter with standard ground joint cone and right angled tube connector.