Erlenmeyerkolvi, vakiohiokset, DURAN®
Toimittaja: DWK Life Sciences
241932602 241935608 241931306 241931709 241931803 241931906 241932405 241932705 241933204 241933607 241933701 241933804 241933907 241934406 241934603 241934706 241932002 241935702 241935402
SCOT241932602EA 260 EUR
SCOT241932602 SCOT241935608 SCOT241931306 SCOT241931709 SCOT241931803 SCOT241931906 SCOT241932405 SCOT241932705 SCOT241933204 SCOT241933607 SCOT241933701 SCOT241933804 SCOT241933907 SCOT241934406 SCOT241934603 SCOT241934706 SCOT241932002 SCOT241935702 SCOT241935402
Erlenmeyerkolvi, vakiohiokset, DURAN®
Kolvit ja erlenmeyerit Erlenmeyerkolvit
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.
According to DIN ISO 4797
*non-DIN ISO size