Bulb pipettes, Class AS, DURAN®

Toimittaja: DWK Life Sciences
233390508 233390051 233390105 233390208 233390302 233390405 233390602 233390705 233390808 233390902 233391007 233391504 233392003 233392509 233393008 233394004 233395009 233390002
SCOT233390508EA 48.4 EUR
SCOT233390508 SCOT233390051 SCOT233390105 SCOT233390208 SCOT233390302 SCOT233390405 SCOT233390602 SCOT233390705 SCOT233390808 SCOT233390902 SCOT233391007 SCOT233391504 SCOT233392003 SCOT233392509 SCOT233393008 SCOT233394004 SCOT233395009 SCOT233390002
Bulb pipettes, Class AS, DURAN®
Pipetit ja pipetintäyttäjät Pallopipetti
Soda-lime glass (AR-Glas®).

  • Class AS
  • Marks and inscription in blue
  • Calibration is based on the poured out volume (Ex)
  • Complies with DIN ISO 648
  • Colour code complies with DIN 12621

Printed Retrace Code (batch identification).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR