Staining jars according to Hellendahl, DURAN®
Toimittaja: DWK Life Sciences
233140001
SCOT233140001EA 305 EUR
SCOT233140001
Staining jars according to Hellendahl, DURAN®
Astiat ja purkit Värjäystölkki
Soda-Lime glass.
For up to 16 microscope slides (76x26 mm).
Varoitus: Note: Do not clean staining dishes and jars at temperatures above 60°C (glass corrosion is possible).