Blancs for reaction vessels, DURAN®
Toimittaja: DWK Life Sciences
210205404 210206306 210207105 210207602 210208607
SCOT210205404EA 49.9 EUR
SCOT210205404 SCOT210206306 SCOT210207105 SCOT210207602 SCOT210208607
Blancs for reaction vessels, DURAN®
Reaktioastiat Reaktioastiat, pullomalli
DURAN® borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Round bottom cylinders.