NS-hiospidikkeet, pidikkeet kartiohioksille, KECK™
Toimittaja: DWK Life Sciences
1090978 1090979 1091142 1091144 1091148 1091140 1091149
SCOT1090978EA 43.7 EUR
SCOT1090978 SCOT1090979 SCOT1091142 SCOT1091144 SCOT1091148 SCOT1091140 SCOT1091149
NS-hiospidikkeet, pidikkeet kartiohioksille, KECK™
Kourat NS-hiospidikkeet
These clips are made from POM to secure glass-to-glass joints.