Rods, AR-Glas®
Toimittaja: Schott AG
1028125 1028126 1028128 1028131 1028133 1028134 1028136 1028140 1028141 1028415 1028425 1028430 1028438
SCOR1028125EA 373 EUR
SCOR1028125 SCOR1028126 SCOR1028128 SCOR1028131 SCOR1028133 SCOR1028134 SCOR1028136 SCOR1028140 SCOR1028141 SCOR1028415 SCOR1028425 SCOR1028430 SCOR1028438
Rods, AR-Glas®
Sauvat
Made of AR®-Glas.
  • Soft glass is acid-resistant
  • Has high thermal expansion for pressure melting and for fusing with platinum or dumet and other alloys