Glass tubings, DURAN® borosilicate glass
Toimittaja: Schott AG
1026144 1027010 1027012 1026014 1027013 1027018 1026134 1026136 1027014 1027016 1027017 1027025 1027011 1026135 1027015 1026141 1027006 1026999 1026125 1027009 1027000 1026142 1026129 1027005 1027001 1027008 1026123 1027007 1026131 1026133 1026127 1026628 1026122 1027021 1027022 1027023 1027024 1027026 1027027 1027028 1026139 1026646 1026718 1027019 1027020 1026138 1139886 1026490 1225058
SCOR1026144EA 1340 EUR
SCOR1026144 SCOR1027010 SCOR1027012 SCOR1026014 SCOR1027013 SCOR1027018 SCOR1026134 SCOR1026136 SCOR1027014 SCOR1027016 SCOR1027017 SCOR1027025 SCOR1027011 SCOR1026135 SCOR1027015 SCOR1026141 SCOR1027006 SCOR1026999 SCOR1026125 SCOR1027009 SCOR1027000 SCOR1026142 SCOR1026129 SCOR1027005 SCOR1027001 SCOR1027008 SCOR1026123 SCOR1027007 SCOR1026131 SCOR1026133 SCOR1026127 SCOR1026628 SCOR1026122 SCOR1027021 SCOR1027022 SCOR1027023 SCOR1027024 SCOR1027026 SCOR1027027 SCOR1027028 SCOR1026139 SCOR1026646 SCOR1026718 SCOR1027019 SCOR1027020 SCOR1026138 SCOR1139886 SCOR1026490 SCOR1225058
Glass tubings, DURAN® borosilicate glass
Letku Siirtoputki
Borosilicate glass acc. to ISO 3585, all-purpose glass for apparatus for the chemical industry, for pipelines and laboratory.
  • High transmittance
  • Electrically highly insulating
  • High chemical resistance
  • High hydrolytic resistance
  • High resistance to thermal shock
  • High temperature resistance
Tube length ~1500 mm.