Ubbelohde viscometers (ASTM)
Toimittaja: SI Analytics
285400567 285400591 285400748 285400756 285400764 285400501 285400518 285400526 285400534 285400542 285400559 285400575 285400583 285400604 285400707 285400715 285400723 285400772 285400789 285400797 285400801 285400731
SCOI285400567EA 539 EUR
SCOI285400567 SCOI285400591 SCOI285400748 SCOI285400756 SCOI285400764 SCOI285400501 SCOI285400518 SCOI285400526 SCOI285400534 SCOI285400542 SCOI285400559 SCOI285400575 SCOI285400583 SCOI285400604 SCOI285400707 SCOI285400715 SCOI285400723 SCOI285400772 SCOI285400789 SCOI285400797 SCOI285400801 SCOI285400731
Ubbelohde viscometers (ASTM)
Viskosimetrit Kapillaariset viskosimetrit
Ubbelohde viscometers for manual measurements of transparent liquids.
Sertifikaatit: ASTM