XYZ Sample changer TW 7450 for piston burettes and automatic titrators
Toimittaja: SI Analytics
285221480 285221400 285221410 285221420 285221430 285221450 285221460 285221470 285221490
SCOI285221480EA 23300 EUR
SCOI285221480 SCOI285221400 SCOI285221410 SCOI285221420 SCOI285221430 SCOI285221450 SCOI285221460 SCOI285221470 SCOI285221490
XYZ Sample changer TW 7450 for piston burettes and automatic titrators
Titraattorit
The Powerful TW 7450 developed for high sample throughput.
  • Very fast and quiet during operation
  • Sample racks with 42, 48 and 72 positions and four different titration heads in different dimensions and holes can be used
  • Titration heads and sample racks can be changed in just a few steps
  • Switch sample racks via software TitriSoft
  • The usable sample vessels have a volume of 50 to 250 ml
  • Connection of one pump for rinsing and suction are possible (diaphragm and peristaltic pump)
  • Additional pumps can be connected via the optional I/O box
  • The calibration of a pH