Viskosimetri, ViscoSystem® AVS® 370 viscosimeter
Toimittaja: SI Analytics
1056509 1056515 1056516 1056517 1056518
616-0028EA 16540 EUR
616-0028 616-0034 SCOI1056516 616-0067 SCOI1056518
Viskosimetri, ViscoSystem® AVS® 370 viscosimeter
Viskosimetrit Automaattiset kapillaariset viskosimetrijärjestelmät
ViscoSystem AVC 370 combines a high degree of measurement accuracy and reproducibility with modularity.
  • Measure by suction (samples with high viscosity) and under pressure (liquids with low boiling points)
  • Modular design: Up to four ViscoPump II modules
  • Multiple measurement station controllable via a PC and multitasking software
  • WinVisco 370 software for two rinsing methods
  • Virtually all standard capillary viscometers can be used
The software allows control of up to two complete AVS 370 systems or a total of eight ViscoPump II modules. The results canMore Product Information