Precision cut thermometers
Toimittaja: Ludwig Schneider GmbH & CO. KG
4736854 4736855 4736856
SCNL4736854EA 89.2 EUR
SCNL4736854 SCNL4736855 SCNL4736856
Precision cut thermometers
Lämpömittarit Analogue Thermometers
These standard grinding size precision cut thermometers are designed with handle.
  • Colourless, round capillary form
Upper part: 275×11 mm
Lower part: Ø 7,25 mm