Ominaispainomittari, tarkat sokerimittarit
Toimittaja: Ludwig Schneider GmbH & CO. KG
3001530 3000015 3007590 3006075 3004560 3003045 3002595 3002523
SCNL3001530EA 130 EUR
SCNL3001530 SCNL3000015 SCNL3007590 SCNL3006075 SCNL3004560 SCNL3003045 SCNL3002595 SCNL3002523
Ominaispainomittari, tarkat sokerimittarit
Ominaispainomittarit
High quality polycarbonate hydrometers for %mas/°Brix measures.
  • Breakage protected (no glass breakage)
  • High transparency with easy-to-read scales
  • High precision and reproducibility
Suitable for applications in food, pharmaceutical and cosmetic industries.
Reference temperature: 20 °C
Tilaustiedot: Available with works calibration certificate on request.
Pakkaus: In square plastic sleeve with individual serial number.