General purpose stirring thermometers
Toimittaja: Ludwig Schneider GmbH & CO. KG
1011701 1011711 1011721 1011731 1011741
SCNL1011701EA 17.9 EUR
SCNL1011701 SCNL1011711 SCNL1011721 SCNL1011731 SCNL1011741
General purpose stirring thermometers
Lämpömittarit Analogue Thermometers
These general purpose stirring thermometers are in stem form.
  • Bulb reinforced by solid glass piece