Incubating orbital shaker, GENIE TEMP-SHAKER 100™
Toimittaja: SCIENTIFIC INDUSTRIES ELMIS
SI-G101 SI-G102
SCIISI-G101EA 3700 EUR
SCIISI-G101 SCIISI-G102
Incubating orbital shaker, GENIE TEMP-SHAKER 100™
Ravistelijat Inkubaatioravistelija
The GENIE TEMP-SHAKER 100™ is designed for gentle orbital shaking, with a speed range from 20 to 100 min⁻¹, while providing precise temperature control from 28 to 75 °C, for use in many laboratory applications.
  • 3-way access clear dome chamber provides 360° view at all times
  • Quick access to samples with easy tilt-n-rest feature using the 'kick stand' feature to open the chamber cover, left, right, or back, to optimise bench space
  • The lightweight cover can also be easily removed for cleaning and attaching accessories
  • Airflow design can heat the chamber to a