Gel filtration chromatography columns, standard, CHROMAFLEX®
Toimittaja: DWK Life Sciences
SP420830-3020 SP420830-1000 SP420830-1520 SP420830-3000 SP420830-6010 SP420830-6020 SP420830-1020 SP420830-1210 SP420830-1220
SCERSP420830-3020EA 1220 EUR
SCERSP420830-3020 SCERSP420830-1000 SCERSP420830-1520 SCERSP420830-3000 SCERSP420830-6010 SCERSP420830-6020 SCERSP420830-1020 SCERSP420830-1210 SCERSP420830-1220
Gel filtration chromatography columns, standard, CHROMAFLEX®
Kromatografiakolonnit
These columns are manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Toimitustiedot: Supplied complete with glass barrel, two end fittings with 20 μm porosity HDPE bed supports, and screw caps. Each column is also supplied with 1,5 m of FEP tubing (1,6 mm OD with 10 mm; 3,2 mm OD with 25 and 48 mm), two flangeless tubing nuts and two flangeless ferrules.