Disposable columns, DISPOSAFLEX®, KIMBLE®
Toimittaja: DWK Life Sciences
SP420160-0000
SCERSP420160-0000EA 452 EUR
SCERSP420160-0000
Disposable columns, DISPOSAFLEX®, KIMBLE®
Kromatografiakolonnit
This is a multi purpose column designed to satisfy a wide range of analytical separations using aqueous buffers and typically used for the extraction of the cyclic nucleotides cAMP and cGMP from sample preparations using AG1-X8.
  • Eliminates the possibility of cross contamination
The lower column fitting with a Luer outlet is molded PP which is designed to secure hold a porous PE bed support or a glass wool plug.