ThermoSafe® AmbiTech® Pre-Qualified 15 to 25 °C Shippers
Toimittaja: Sonoco Thermosafe
ATG22 ATG65 ATR28
SCATATG22EA 1820 EUR
SCATATG22 SCATATG65 SCATATR28
ThermoSafe® AmbiTech® Pre-Qualified 15 to 25 °C Shippers
Lähetys- ja kuljetuslaatikot
AmbiTech® pre-qualified shippers use an inorganic salt-based PCM to provide exceptional temperature control.
  • Pre-qualified 15 to 25 °C shipper
  • Tested to several ambient profiles to speed up implementation.
  • Universal packout simplifies operations; no need to manage the switch from summer to winter and back again
  • Integrated handles for easy movement
AmbiTech® controlled room temperature systems use PCM to deliver superior temperature control for temperature-sensitive pharmaceuticals, even in the harshest of climates. These systems incorporate AmbiSorb®,More Product Information