Nalidixic acid sodium salt

Toimittaja: SCHARLAB

6124LYO 06-124-CASE
SCAR6124LYOEA 0 EUR
SCAR6124LYO SCAR06-124-CASE
Nalidixic acid sodium salt
Nalidixic acid sodium salt
Kaava: C₁₂H₁₁N₂NaO₃
MDL numero: MFCD00064376
CAS-numero: 3374-05-8
EINECS: 222-159-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR