Sample Bags without Closure
Toimittaja: SCANCELL FOLIEN VETRIEB
2066_U 2047 2046 2077 2078 10034060 2092 12001865 2090 2080 20771 2063
129-9127EA 15.8 EUR
129-9127 SCAC2047 SCAC2046 SCAC2077 SCAC2078 SCAC10034060 SCAC2092 SCAC12001865 SCAC2090 SCAC2080 SCAC20771 SCAC2063
Sample Bags without Closure
Pussit ja sulkijat Näytepussit
Bags made from PE transparent, for the storage or transport of samples.
Available in several sizes.
Varoitus: Supplied without closure ties.