Tork Hand Sanitising Alcohol Foam
Toimittaja: Essity
590102
SCAA590102EA 192 EUR
SCAA590102
Tork Hand Sanitising Alcohol Foam
Käsienpesu- ja desinfektioaineet, hoitovoiteet Desinfektioaineet käsille Liquid and Foam Skin Disinfectants
Tork hand sanitising alcohol foam, gentle to the skin.
  • Conform with EN 1500 and EN 14476 (all enveloped, such as HIV, SARS Corona, HBV, HCV, Influenza H1N1 and H5N1) with this effective formula
  • Provide a hand sanitiser that's gentle to the skin and easy to handle - this foam formula spreads easily
  • Show that you care for user's skin, thanks to this alcohol hand sanitiser's dermatologically tested formula
Help prevent the spread of infections with Tork hand sanitising alcohol foam. Containing 75% w/w denatured alcohol, it is ideal for sanitising hands