Lentivirus purification kits, Vivapure® LentiSELECT
Toimittaja: Sartorius
VS-LVPQ040 VS-LVPQ1000 VS-LVPQ500
SARTVS-LVPQ040EA 986 EUR
SARTVS-LVPQ040 SARTVS-LVPQ1000 SARTVS-LVPQ500
Lentivirus purification kits, Vivapure® LentiSELECT
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
LentiSELECT kits offer LentiSELECT 40, LentiSELECT 500 and LentiSELECT 1000 for the purification and concentration of VSV-G pseudotyped lentivirus from 40 to 1000 ml cell culture, leading to purified infective particles. These lentivirus purification and concentration kits offer purified recombinant lentivirus particles for in-vitro transfection a fast, safe and easy to use solution. These kits replace time-consuming ultracentrifugation protocols, which typically take about one day for large sample volumes, thus reducing the purification time to only a few hours.
  • High virus purity
  • Fast and easy virus purification
  • Low endotoxin levels and high cell viability
  • Optimal for multiple virus construct screening
  • Independent of other equipment such as ultracentrifuges
Toimitustiedot: Each kit consist of LentiSELECT units, syringe, tube set with one-way valve, loading buffer (10×), washing buffer, elution buffer, Vivaspin 20 and operating manual (each for kit and Vivaspin).