Quartz microfibre filters, pre-heated, grade Q3400
Toimittaja: Sartorius
Q3400-20-G Q3400-25-G Q3400-37-G Q3400-45-G Q3400-47-G Q3400-50-G Q3400-82-N Q3400-90-N Q3400-142-K Q3400-150-K
SARTQ3400-20-GEA 77.7 EUR
SARTQ3400-20-G SARTQ3400-25-G SARTQ3400-37-G SARTQ3400-45-G SARTQ3400-47-G SARTQ3400-50-G SARTQ3400-82-N SARTQ3400-90-N SARTQ3400-142-K SARTQ3400-150-K
Quartz microfibre filters, pre-heated, grade Q3400
Suodattimet Lasikuitusuodattimet
These 100% high-purity quartz microfibre filters are free of glass fibres and binding agents. They are especially suited for emission monitoring at temperature of up to 900 °C and wherever filters of the highest purity are needed.
  • Highly temperature and chemical resistant
  • Excellent weight and dimensional stability
  • Biologically inert
Tested and classified according to the standard EN 143.