Qualitative filter papers, technical, smooth, grade C 140
Toimittaja: Sartorius
FT-4-356-150 FT-4-356-110 FT-4-356-240 FT-4-356-185 FT-3-356-090
SARTFT-4-356-150EA 321 EUR
SARTFT-4-356-150 SARTFT-4-356-110 SARTFT-4-356-240 SARTFT-4-356-185 SARTFT-3-356-090
Qualitative filter papers, technical, smooth, grade C 140
Suodattimet Suodatinpaperit Kvalitatiivinen suodatinpaperi
These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%.
They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Medium fast rate of filtration.