Qualitative filter papers, technical, creped
Toimittaja: Sartorius
FT-2-483580580 FT-2-342580580 FT-4-423-125 FT-3-423-090 FT2-335-580580 FT-3-423-240 FT-3-335-125 FT-3-423-185 FT-3-423-150 FT-3-335-270 FT-3-335-150 FT-3-335-110 FT-3-335-185 FT-4-423-185 FT-4-423-385 FT-4-423-320 FT-4-423-240 FT-4-423-150 FT-4-423-270 FT-3-314-150 FT-3-314-090 FT-4-423-110 FT-2-354580580 FT-2-423580580 FT-4-483-185 FT-4-480-150 FT-2-314580580 FT-4-483-500 FT-3-342-125 FT-3-354-185 FT-3-354-240 FT-3-354-320 FT-3-423-110 FT-3-423-125 FT-4-483-240 FT-3-480-185 FT-3-314-110 FT-3-480-125 FT-3-335-090 FT-3-335-190 FT-3-335-200 FT-3-480-150 FT-3-314-070 FT-3-314-240 FT-3-314-385 FT-4-335-125 FT-4-335-240 FT-4-335-320 FT-4-423-500 FT2-480-580580
515-1613EA 587 EUR
515-1613 515-1623 SARTFT-4-423-125 SARTFT-3-423-090 512-0536 SARTFT-3-423-240 SARTFT-3-335-125 SARTFT-3-423-185 SARTFT-3-423-150 SARTFT-3-335-270 SARTFT-3-335-150 SARTFT-3-335-110 SARTFT-3-335-185 SARTFT-4-423-185 SARTFT-4-423-385 SARTFT-4-423-320 SARTFT-4-423-240 SARTFT-4-423-150 SARTFT-4-423-270 SARTFT-3-314-150 SARTFT-3-314-090 SARTFT-4-423-110 SARTFT-2-354580580 SARTFT-2-423580580 SARTFT-4-483-185 SARTFT-4-480-150 SARTFT-2-314580580 SARTFT-4-483-500 SARTFT-3-342-125 SARTFT-3-354-185 SARTFT-3-354-240 SARTFT-3-354-320 SARTFT-3-423-110 SARTFT-3-423-125 SARTFT-4-483-240 SARTFT-3-480-185 SARTFT-3-314-110 SARTFT-3-480-125 SARTFT-3-335-090 SARTFT-3-335-190 SARTFT-3-335-200 SARTFT-3-480-150 SARTFT-3-314-070 SARTFT-3-314-240 SARTFT-3-314-385 SARTFT-4-335-125 SARTFT-4-335-240 SARTFT-4-335-320 SARTFT-4-423-500 SARTFT2-480-580580
Qualitative filter papers, technical, creped
Suodattimet Suodatinpaperit Kvalitatiivinen suodatinpaperi
These filter papers are used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore
suitable for pressure or vacuum filtration.
They are made of refined pulp and linters with an >95% alpha-cellulose content, are very pure with an ash content between <0,1 to 0,15%.
Grade 5 H/N: Very fast rate of filtration, wide-pore. Used for filtration of essential oils.
Grade 37/N: Very fast rate of filtration, wide-More Product Information