Glass microfibre filters without binder
Toimittaja: Sartorius
13440-130-K 13440-44-Q 13440-50-Q 13440-100-K FT-3-1101-125 FT-3-1105-025 FT-3-1105-050 FT-3-1101-055 FT-3-1103-047 FT-3-1103-025 FT-3-1101-150 FT-3-1101-110 FT-3-1101-070 FT-3-1103-100 FT-3-1101-047 FT-3-1101-025 FT-3-1101-090 FT-3-1105-125 FT-3-1103-055 FT-3-1103-090 FT-3-1105-110 FT-3-1105-090 FT-3-1105-055 FT-3-1105-047 FT-3-1104-025 FT-3-1103-150 FT-3-1103-125 FT-3-1103-110 13440-47-Q 13440-42-Q FT-3-1102-110 FT-3-1102-150 FT-3-1101-080 FT-3-1102-090 FT-3-1104-013 FT-3-1104-047 FT-3-01110-090 FT-3-01110-110 FT-3-1101-050 FT-3-1101-185 FT-3-1101-240 FT-3-1102-025 FT-3-1102-047 FT-3-1102-050 FT-3-1102-070 FT-3-1102-100 FT-3-1102-125 FT-3-1103-050 FT-3-1103-070 FT-3-1103-185 FT-3-1104-050 FT-3-1104-070 FT-3-1104-090 FT-3-1104-100 FT-3-1104-110 FT-3-1104-150 FT-3-1105-070 FT-3-01110-050 FT-3-1106-047 FT-3-1106-055 FT-3-1106-125 FT-3-1105-142 FT-3-1106-025 FT-3-1106-090 FT-3-1106-110 FT-3-1104-045 FT-3-1106-070 FT-3-01110-070 FT-3-1101-037 FT-3-1103-142 FT-3-01147-090 FT-3-1106-037
SART13440-130-KEA 238 EUR
SART13440-130-K SART13440-44-Q 512-0085 SART13440-100-K 516-7103 513-0076 516-7117 516-7098 516-7109 516-7108 516-7104 516-7102 516-7099 516-7112 516-7097 516-7096 516-7101 516-7120 516-7110 516-7111 516-7119 513-0078 516-7118 513-0077 516-7116 516-7115 516-7114 516-7113 SART13440-47-Q SART13440-42-Q 516-7106 516-7107 SARTFT-3-1101-080 516-7105 SARTFT-3-1104-013 SARTFT-3-1104-047 SARTFT-3-01110-090 SARTFT-3-01110-110 SARTFT-3-1101-050 SARTFT-3-1101-185 SARTFT-3-1101-240 513-0095 SARTFT-3-1102-047 SARTFT-3-1102-050 SARTFT-3-1102-070 SARTFT-3-1102-100 SARTFT-3-1102-125 SARTFT-3-1103-050 513-0096 SARTFT-3-1103-185 SARTFT-3-1104-050 SARTFT-3-1104-070 SARTFT-3-1104-090 SARTFT-3-1104-100 SARTFT-3-1104-110 SARTFT-3-1104-150 SARTFT-3-1105-070 SARTFT-3-01110-050 SARTFT-3-1106-047 SARTFT-3-1106-055 SARTFT-3-1106-125 SARTFT-3-1105-142 SARTFT-3-1106-025 SARTFT-3-1106-090 SARTFT-3-1106-110 SARTFT-3-1104-045 SARTFT-3-1106-070 SARTFT-3-01110-070 SARTFT-3-1101-037 SARTFT-3-1103-142 SARTFT-3-01147-090 SARTFT-3-1106-037
Glass microfibre filters without binder
Suodattimet Lasikuitusuodattimet
Binder free glass microfibre filters are made of 100% borosilicate, are highly pure and extremely white. They are recommended for analytical and gravimetric analyses and also as prefilters. These filters combine fast flow rates with high load capacity and the retention of very fine particles.
  • Biologically inert
  • Chemical resistant
Grade MGA: Clarification of buffer and reagent solutions
Grade MG 160: Air monitoring
Grade MG 550-HA : Analysis of suspended solids in wastewater according to 2540D
Grade MGB, Grade MGD, Grade 13440: Membrane prefilter