Ruiskusuodatin Minisart® NY
Toimittaja: Sartorius
17845-ACK 17845-Q 17846-ACK 17846-Q 1784B-K 1784B-Q 1784C-Q 1784C-K 1776B-K 1776B-Q 1776C-K 1776C-Q 17846-R 17845-R
514-0080EA 235 EUR
514-0080 514-0081 514-0082 514-0083 SART1784B-K SART1784B-Q SART1784C-Q 516-0684 SART1776B-K SART1776B-Q 516-0681 SART1776C-Q SART17846-R SART17845-R
Ruiskusuodatin Minisart® NY
Suodattimet Ruiskusuodattimet
Minisart® NY with hydrophilic polyamide (nylon) is optimised for aqueous liquids and solvents with a pH of 3 to 14. Type 17845 with a pore size of 0,2 µm can be used for sample preparation and sterile filtration. It has a male Luer slip outlet and version ACK is individually packed and sterilised by ethylene oxide (EO).
  • Sterile and ready-to-use
  • Minimum extractables or leachables
  • Low non-specific binding
  • Outperforming chemical compatibility
Individually packed and sterile Minisart® NY syringe filters are optimal for immediate usage whenever sterileMore Product Information