Ruiskusuodatin, Minisart® aqueous filtration
Toimittaja: Sartorius
1776D-ACK 1757H-K 1757H-Q 1776D-Q
516-0682EA 261 EUR
516-0682 SART1757H-K SART1757H-Q SART1776D-Q
Ruiskusuodatin, Minisart® aqueous filtration
Suodattimet Ruiskusuodattimet
Minisart® PES with polyethersulfone (PES) is optimal for highest flow rates and a pH of 2 - 13. Type 1776D with a pore size of 0,22 µm can be used for sterile filtration and has a male Luer slip outlet. Version ACK is individually packed and sterilised by ethylene oxide (EO).
  • Sterile and ready-to-use
  • Slightly asymmetric PES with superior flow rate
  • Low non-specific binding
  • Bidirectional use possible
Individually packed and sterile Minisart® PES syringe filters are optimal for immediate usage whenever sterile filtration or clarification and cell removalMore Product Information