Sterility test system, Sterisart® NF
Toimittaja: Sartorius
16466-GBD 16467-GBD 16468-GBD 16469-GBD 16470-GBD 16475-GBD 16476-GBD 16477-GBD 16478-GBD 16466-GSD 16467-GSD 16475-GSD 16476-GSD 16469-GSD
SART16466-GBDEA 759 EUR
SART16466-GBD SART16467-GBD SART16468-GBD SART16469-GBD SART16470-GBD SART16475-GBD SART16476-GBD SART16477-GBD SART16478-GBD SART16466-GSD SART16467-GSD SART16475-GSD SART16476-GSD SART16469-GSD
Sterility test system, Sterisart® NF
Suodattimet Membraanisuodatinsuppilot
The Sterisart® NF System is a completely disposable technology based on the membrane filtration method, for the sterility testing of pharmaceutical products. It makes aseptic sampling easy, during or after incubation which is required for dilution or rapid detection, or for the supplement of additives for inactivation of antibiotics.
  • Sterisart NF alpha, dual-needle metal spike for closed containers
  • Sharp Metal needle for easy penetration
  • High retention of microbes
  • Low adsorption
  • Huge variety of adapters and more than 20 different unit types
  • Gas-impermeable