Sterisart® Universal, Gen 4 Peristaltic Sterility Testing Pumps
Toimittaja: Sartorius
16421-A 16421-B 16421-E
SART16421-AEA 43600 EUR
SART16421-A SART16421-B SART16421-E
Sterisart® Universal, Gen 4 Peristaltic Sterility Testing Pumps
Pumput Peristalttiset pumput
The Sterisart® Universal pump, Gen 4 is a robust and reliable peristaltic sterility testing pump built from 316L mirror-finish stainless steel and used for sample and media transfer into the Sterisart® family of sterility testing canisters.
  • Low and compact space-saving profile
  • Equal splitting of liquid volumes
  • 316L Grade mirror-finish stainless steel
  • Robust and durable: Compatible with all commonly used sterilants and fully VHP resistant
  • Open design: Compatible with other filtration devices
  • Real-time pressure sensor
  • Integrated barcode scanner supporting 28