Ultrafiltration membrane filters, cellulose triacetate
Toimittaja: Sartorius
14529-47-D 14539-47-D 14549-47-D 14549-47-N 14529-25-D 14539-25-D 14549-25-D 14549-63-D 14549-43-D 14539-50-D
SART14529-47-DEA 182 EUR
SART14529-47-D SART14539-47-D SART14549-47-D SART14549-47-N SART14529-25-D SART14539-25-D SART14549-25-D SART14549-63-D SART14549-43-D SART14539-50-D
Ultrafiltration membrane filters, cellulose triacetate
Suodattimet Membraanisuodattimet
These membrane filters have high hydrophilicity and exceptionally low non-specific binding characteristics. These membranes are ideal for sample cleaning and protein removal and when high recovery of the filtrate solution is of primary importance.
  • Cast without any membrane support that could trap or bind passing microsolutes