Nutrient Pad Sets, NKS e.motion
Toimittaja: Sartorius
14055-50-RDN 14080-50-RDN 14092-50-RDN 14058-50-RDN 14058-50-RKN 14055-47-RDN 14062-47-RDN 14092-47-RDN 14051-47-RDN 14075-47-RDN 14091-47-RDN 14084-47-RDN 14087-47-RDN 14087-50-RDN 14076-50-RDN 14094-50-RDN 14096-47-RDN 14096-50-RDN 14091-50-RDN 14051-50-RDN 14053-50-RDN 14058-47-RDN 14062-50-RDN 14075-50-RDN 14076-47-RDN 14080-47-RDN 14056-47-RDN
SART14055-50-RDNEA 297 EUR
SART14055-50-RDN SART14080-50-RDN SART14092-50-RDN SART14058-50-RDN SART14058-50-RKN SART14055-47-RDN SART14062-47-RDN SART14092-47-RDN SART14051-47-RDN SART14075-47-RDN SART14091-47-RDN SART14084-47-RDN SART14087-47-RDN SART14087-50-RDN SART14076-50-RDN SART14094-50-RDN SART14096-47-RDN SART14096-50-RDN SART14091-50-RDN SART14051-50-RDN SART14053-50-RDN SART14058-47-RDN SART14062-50-RDN SART14075-50-RDN SART14076-47-RDN SART14080-47-RDN SART14056-47-RDN
Nutrient Pad Sets, NKS e.motion
Elatusaineet mikrobiologiaan Valmisteltu viljelyaine
Nutrient pad sets for colony counting by the membrane filter method, specially designed for the Microsart e.motion dispenser. These nutrient pad consist of dehydrated culture media inserted into pre-sterilised Petri dishes. This alternative to agar media has a long storage life of 24 months at room temperature.
  • Economical, time-saving microbiological control
  • High quality and reproducible results for up to 24 months
  • Compliant with international standards and references
  • Easy handling