Racks for tubes and S-Monovettes, PP
Toimittaja: SARSTEDT
93.854.122 93.852.171 93.854.125 93.851 93.851.171 93.851.172 93.851.173 93.851.174 93.851.175 93.852 93.852.168 93.852.169 93.852.170 93.852.172 93.852.173 93.852.174 93.852.175 93.852.200 93.852.300 93.853 93.853.131 93.853.134 93.853.135 93.853.200 93.853.300 93.854 93.854.121 93.854.123 93.854.124
SARS93.854.122EA 24.3 EUR
SARS93.854.122 SARS93.852.171 SARS93.854.125 SARS93.851 SARS93.851.171 SARS93.851.172 SARS93.851.173 SARS93.851.174 SARS93.851.175 SARS93.852 SARS93.852.168 SARS93.852.169 SARS93.852.170 SARS93.852.172 SARS93.852.173 SARS93.852.174 SARS93.852.175 SARS93.852.200 SARS93.852.300 SARS93.853 SARS93.853.131 SARS93.853.134 SARS93.853.135 SARS93.853.200 SARS93.853.300 SARS93.854 SARS93.854.121 SARS93.854.123 SARS93.854.124
Racks for tubes and S-Monovettes, PP
Koeputkitelineet Tube Racks
Made of PP.
  • Autoclavable up to 121 °C
  • Suitable for space-saving sample storage
Universal block racks for wide range of applications and available in different sizes and colours.