Tissue culture flasks
Toimittaja: SARSTEDT
83.1812 83.1813 83.3911.002 83.1810 83.1810.001 83.1811.001 83.1811.002 83.1812.001 83.1812.002 83.1813.001 83.3910.002
SARS83.1812EA 0 EUR
SARS83.1812 SARS83.1813 SARS83.3911.002 SARS83.1810 SARS83.1810.001 SARS83.1811.001 SARS83.1811.002 SARS83.1812.001 SARS83.1812.002 SARS83.1813.001 SARS83.3910.002
Tissue culture flasks
Kolvit ja erlenmeyerit Soluviljelypullot
High-quality polystyrene.
  • Sterilised by gamma irradiation
  • Hydrophilic surface for optimal attachment and growth of adherent cells
  • Anti-tip skirt at canted neck
With red PE quick release (with or without ventilation) screw cap or black PP rim seal cap.