Sample cups for Cobas system
Toimittaja: SARSTEDT
73.911
SARS73.911EA 0 EUR
SARS73.911
Sample cups for Cobas system
Näyteastiat, muovia Sample Cups for Analysers
Sample cups for Cobas system such as Bio, Mira, Fara and Integra.
  • Good chemical resistance
  • Good temperature resistance
Cups for Integra has cap with hole to insert its probe.