PCR tubes, Multiply®
Toimittaja: SARSTEDT
72.737.002 72.735.002 72.735.992 72.737.992 72.985.002 72.991.002 72.735.100 72.737 72.985.992 72.990 72.990.002 72.990.992 72.991.992
SARS72.737.002EA 249 EUR
SARS72.737.002 SARS72.735.002 SARS72.735.992 SARS72.737.992 SARS72.985.002 SARS72.991.002 SARS72.735.100 SARS72.737 SARS72.985.992 SARS72.990 SARS72.990.002 SARS72.990.992 SARS72.991.992
PCR tubes, Multiply®
Putket PCR-putket
PP, translucent
  • Thin, smooth walls ensure rapid and even heat transfer to the sample
  • Anti-contamination shield prevents accidental contact with the inner surface of the lid, preventing potential contamination
  • Frosted flat lid with large writing area
Developed due to the limitations of traditional reaction tubes they offer many desirable benefits. Enhanced design features ensure work is risk-free from contamination and non-tiring compared to the more traditional tubes. 0,2 and 0,5 ml thin-walled Multiply® PCR cups are moulded with an integral anti-More Product Information