Microcentrifuge tubes, with or without attached caps
Toimittaja: SARSTEDT
72.690.476 72.690 72.690.007 72.690.475 72.690.477 72.690.478 72.696 72.708 72.689 72.688 72.690.001 72.690.550 72.709 72.688.001 72.688.002 72.688.003 72.688.004 72.688.005 72.689.001 72.689.002 72.689.003 72.689.004 72.689.006 72.690.004 72.698 72.698.200 72.699 72.699.001 72.699.002 72.699.003 72.699.004 72.699.005 72.691
SARS72.690.476EA 0 EUR
SARS72.690.476 SARS72.690 SARS72.690.007 SARS72.690.475 SARS72.690.477 SARS72.690.478 SARS72.696 SARS72.708 SARS72.689 SARS72.688 SARS72.690.001 SARS72.690.550 SARS72.709 SARS72.688.001 SARS72.688.002 SARS72.688.003 SARS72.688.004 SARS72.688.005 SARS72.689.001 SARS72.689.002 SARS72.689.003 SARS72.689.004 SARS72.689.006 SARS72.690.004 SARS72.698 SARS72.698.200 SARS72.699 SARS72.699.001 SARS72.699.002 SARS72.699.003 SARS72.699.004 SARS72.699.005 SARS72.691
Microcentrifuge tubes, with or without attached caps
Putket Mikrosentrifugiputket
PP, translucent.
  • Autoclavable
  • Good chemical resistance
  • Non sterile
For transport, storage and sample processing in the laboratory.