Microtubes, with thread
Toimittaja: SARSTEDT
72.694.005 72.730.006 72.692 72.693 72.693.005 72.609 72.692.005 72.694.006 72.607 72.608 72.609.003 72.609.001 72.664 72.687 72.703 72.703.004 72.730 72.730.003 72.730.004 72.730.007 72.730.009 72.733.001 72.733.010 72.785.005
SARS72.694.005EA 241 EUR
SARS72.694.005 SARS72.730.006 SARS72.692 SARS72.693 SARS72.693.005 SARS72.609 SARS72.692.005 SARS72.694.006 SARS72.607 SARS72.608 SARS72.609.003 SARS72.609.001 SARS72.664 SARS72.687 SARS72.703 SARS72.703.004 SARS72.730 SARS72.730.003 SARS72.730.004 SARS72.730.007 SARS72.730.009 SARS72.733.001 SARS72.733.010 SARS72.785.005
Microtubes, with thread
Putket Klusteriputket
PP, translucent.
  • Safe screw cap with O-ring
  • Sterile or non sterile
  • Autoclavable
For transport, storage and sample processing in the laboratory.