Microtubes with thread in special sterile packaging
Toimittaja: SARSTEDT
72.607.772 72.609.711 72.664.711 72.687.772 72.730.711
SARS72.607.772EA 302 EUR
SARS72.607.772 SARS72.609.711 SARS72.664.711 SARS72.687.772 SARS72.730.711
Microtubes with thread in special sterile packaging
Putket Klusteriputket
PP, translucent
  • With or without skirted base
Each bag is sub-packaged into a second additional bag. Removing the outer bag leaves a complete and sterile bag packaging for convenient processing in clean rooms or sterile environments. This packaging option is ideal for users working under clean room conditions to fill precious reagents into tubes.