Microtubes, prepared
Toimittaja: SARSTEDT
41.1378.005 41.1505.005 41.1503.005 41.1504.005 41.1395.005 41.1500.005 41.1350.005 41.1392.005 41.1393.005 41.1394.005 41.1501.005 41.1506 41.1506.002 41.1506.005
SARS41.1378.005EA 560 EUR
SARS41.1378.005 SARS41.1505.005 SARS41.1503.005 SARS41.1504.005 SARS41.1395.005 SARS41.1500.005 SARS41.1350.005 SARS41.1392.005 SARS41.1393.005 SARS41.1394.005 SARS41.1501.005 SARS41.1506 SARS41.1506.002 SARS41.1506.005
Microtubes, prepared
Putket Klusteriputket
PP, translucent with PP screw cap.
  • Type of preparation: Trisodium citrate solution
  • With paper label
  • Graduated
  • Recommended centrifugation: 1800×g
With screw cap or push-on cap.