Blood collecting tubes with serum gel (clotting activator), S-Monovette®
Toimittaja: SARSTEDT
06.1667.001 04.1935 01.1602.001 03.1524.001 04.1905.001 04.1935.001 02.1388.001 01.1602.00 02.1388 03.1524 04.1905 04.1905.001-50 04.1935.001-50 01.1602.001_50
SARS06.1667.001EA 299 EUR
SARS06.1667.001 SARS04.1935 SARS01.1602.001 SARS03.1524.001 SARS04.1905.001 SARS04.1935.001 SARS02.1388.001 SARS01.1602.00 SARS02.1388 SARS03.1524 SARS04.1905 SARS04.1905.001-50 SARS04.1935.001-50 SARS01.1602.001_50
Blood collecting tubes with serum gel (clotting activator), S-Monovette®
Diagnostiset testit Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
S-Monovette® is a blood collection system that combines the aspiration and the vacuum techniques.
  • Aspiration technique for routine blood collection
  • Vacuum technique is always available
  • For all ages, from young to old
Suitable for all vein conditions and achieves a sample quality for high quality results.